Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927375 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006719904.3 | XP_006719967.1 | 1 | 170 | ||
XM_011535334.2 | XP_011533636.1 | 1 | 170 | |||
XM_011535335.2 | XP_011533637.1 | 1 | 170 | |||
XM_011535336.2 | XP_011533638.1 | 1 | 170 |
Gene Symbol: ANKRD26P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027248.3 | 1 | 83 | |||
GenBank mRNA | BC036210.1 | - | 1 | 83 |