Gene Symbol: VTI1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006717637.2 | XP_006717700.1 | 9 | 1736 | ||
XM_011539331.2 | XP_011537633.1 | 9 | 1747 | |||
XM_017015748.1 | XP_016871237.1 | 8 | 1669 |
Gene Symbol: LOC143188 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK026759.1 | - | 1 | 627 | ||
AK074471.1 | - | 2 | 2182 |