Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARGP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029388.2 | 11, 12 | 1255 | |||
GenBank mRNA | AK293897.1 | - | 7, 8 | 783 | ||
AK311609.1 | - | 11, 12 | 1227 |