Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLJ37201 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026835.1 | 2 | 350 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000455699.1 | - | 2 | 350 | Chr. 10 - 89694295 - 89697928 [-] on Build GRCh38 |