Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00339 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023918.2 | 2 | 343 | |||
NR_023919.2 | 2 | 301 | ||||
NR_109759.1 | 2 | 297 | ||||
NR_109760.1 | 2 | 298 | ||||
NR_109761.1 | 3 | 418 | ||||
NR_109762.1 | 3 | 422 | ||||
GenBank mRNA | AF161506.1 | - | 2 | 271 | ||
BC016053.1 | - | 2 | 256 | |||
BG432520.1 | - | 3 | 412 | |||
BX509008.1 | - | 2 | 298 | |||
CA489192.1 | - | 2 | 320 |