Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMIZ1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015429.1 | 6 | 572 | |||
NR_024429.1 | 6 | 668 | ||||
NR_024431.2 | 8 | 864 | ||||
GenBank mRNA | AK097923.1 | - | 1 | 1072 | ||
AK098249.1 | - | 8 | 1309 | |||
BC102000.1 | - | 5 | 415 | |||
BC102001.1 | - | 5 | 415 | |||
BC105703.1 | - | 5 | 511 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA541438.1 | - | 6 | 554 | Chr. 10 - 78961290 - 79068531 [-] on Build GRCh38 |