Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RN7SK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_001445.2 | 1 | 190 | |||
GenBank mRNA | X04236.1 | - | LIT1704 7SK |
1 | 189 |
Gene Symbol: PLCG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002661.4 | NP_002652.2 | 33 | 8129 | ||
GenBank mRNA | AK127374.1 | - | 1 | 1504 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK058127.1 | - | 190 | Chr. - - [-] on Build |