Gene Symbol: C21orf62-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024622.1 | 4 | 1107 | |||
NR_024623.1 | 4 | 1095 | ||||
GenBank mRNA | AK096601.1 | - | 4 | 1096 | ||
BC034794.1 | - | 2 | 821 | |||
BC117397.1 | - | 4 | 989 | |||
BC117399.1 | - | 4 | 1001 |