Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR7E156P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002171.2 | 1 | 588 | |||
GenBank mRNA | AK097469.1 | - | 1 | 588 | ||
AK097490.1 | - | 2 | 1198 | |||
AK097573.1 | - | 2 | 1237 |
Gene Symbol: LOC102723968 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130763.1 | 1 | 551 | |||
GenBank mRNA | AK098560.1 | - | 1 | 551 | ||
AK127969.1 | - | 2 | 1676 |