Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: HCG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002139.2 | 1 | 1701 | |||
GenBank mRNA | AF036977.1 | - | 1 | 1699 | ||
BC011879.1 | - | 1 | 1485 |