Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033262.1 | 1 | 121 | |||
GenBank mRNA | AK074574.1 | - | 1 | 121 | ||
BC128256.1 | - | 1 | 65 | 40115869 | ||
BC128409.1 | - | 1 | 65 | 40115871 |