Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY6G6E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003673.3 | 1 | 13 | |||
NR_024541.1 | 1 | 13 | ||||
GenBank mRNA | DA786728.1 | - | 1 | 13 |