Search Thermo Fisher Scientific
Gene Symbol: LINC00934 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024246.2 | 4 | 1704 | |||
GenBank mRNA | AK057497.1 | - | 4 | 1704 | ||
BC101457.1 | - | 1 | 707 | |||
BC101458.1 | - | 1 | 707 | |||
BC101459.1 | - | 1 | 707 | |||
BC118554.1 | - | 1 | 707 | |||
BC118632.1 | - | 2 | 621 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000630357.1 | - | 1704 | Chr. - - [-] on Build |