Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029679.1 | 1 | 58 |
Gene Symbol: MIR137HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046105.1 | 3 | 708 | |||
GenBank mRNA | AK094607.1 | - | 3 | 620 | ||
AK309618.1 | - | 1 | 108 | |||
AK311400.1 | - | 1 | 111 |