Search Thermo Fisher Scientific
Gene Symbol: LOC202181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026921.1 | 1, 2 | 262 |
Gene Symbol: SIMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308195.1 | NP_001295124.1 | 1, 2 | 284 | ||
GenBank mRNA | AK299336.1 | - | 1, 2 | 244 | ||
DC372557.1 | - | 1, 2 | 244 | |||
KF151170.1 | - | 1, 2 | 152 |