Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01554 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026936.1 | 3 | 1191 | |||
GenBank mRNA | AK096140.1 | - | 4 | 1090 | ||
AY168789.1 | - | 4 | 1070 |