Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC37A4P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002940.2 | 2, 3 | 2766 | |||
GenBank mRNA | AK000982.1 | - | 2, 3 | 895 |