Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCARNA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002998.1 | 1 | 93 |
Gene Symbol: RFWD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017002059.1 | XP_016857548.1 | 20 | 6121 | ||
XM_017002060.1 | XP_016857549.1 | 20 | 6109 | |||
XM_017002061.1 | XP_016857550.1 | 19 | 6067 | |||
XM_017002062.1 | XP_016857551.1 | 19 | 6061 | |||
XM_017002063.1 | XP_016857552.1 | 19 | 6055 | |||
XM_017002064.1 | XP_016857553.1 | 19 | 6052 | |||
XM_017002065.1 | XP_016857554.1 | 18 | 6007 | |||
XM_017002066.1 | XP_016857555.1 | 18 | 5998 | |||
XM_017002067.1 | XP_016857556.1 | 18 | 5995 | |||
XM_017002068.1 | XP_016857557.1 | 18 | 5992 | |||
XM_017002069.1 | XP_016857558.1 | 14 | 5635 | |||
XM_017002070.1 | XP_016857559.1 | 19 | 5827 | |||
XM_017002071.1 | XP_016857560.1 | 17 | 5216 | |||
XM_017002072.1 | XP_016857561.1 | 17 | 5214 | |||
XM_017002073.1 | XP_016857562.1 | 16 | 5156 | |||
XM_017002074.1 | XP_016857563.1 | 16 | 5154 | |||
XM_017002075.1 | XP_016857564.1 | 16 | 5236 | |||
XM_017002077.1 | XP_016857566.1 | 15 | 5011 | |||
XM_017002078.1 | XP_016857567.1 | 17 | 5284 | |||
XM_017002079.1 | XP_016857568.1 | 17 | 5699 | |||
XM_017002080.1 | XP_016857569.1 | 13 | 4938 |