Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNORA62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002324.1 | 1 | 51 | |||
GenBank mRNA | L07383.1 | - | 1 | 51 |
Gene Symbol: RPSA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK055991.1 | - | 1 | 846 |