Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM41C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027055.1 | 3 | 1190 |
Gene Symbol: LOC101927506 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017014134.1 | XP_016869623.1 | 4 | 2290 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG215690.1 | - | 1 | 94 | Chr. 1 - 868071 - 868675 [-] on Build GRCh38 |