Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FLJ13224 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026806.1 | 1 | 1151 | |||
GenBank mRNA | AK023286.1 | - | 1 | 1151 | ||
BC118557.1 | - | 1 | 978 | |||
BC118637.1 | - | 1 | 978 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX107136.1 | - | 1 | 44 | Chr. 12 - 31325423 - 31325817 [+] on Build GRCh38 |