Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLJ13224 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026806.1 | 1 | 728 | |||
GenBank mRNA | AK023286.1 | - | 1 | 728 | ||
BC118557.1 | - | 1 | 555 | |||
BC118637.1 | - | 1 | 555 |