Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HERC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022697.1 | XP_016878186.1 | 1 | 210 |
Gene Symbol: HERC2P4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC038215.2 | - | 2 | 559 |