Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC145474 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027046.1 | 1 | 1650 | |||
GenBank mRNA | AK023718.1 | - | 1 | 1650 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DW418627.1 | - | 1 | 367 | Chr. 14 - 71489160 - 71489563 [+] on Build GRCh38 |