Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D3P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027486.1 | 8 | 571 | |||
GenBank mRNA | AK301746.1 | - | 8 | 572 | ||
BC058890.1 | - | 8 | 596 |