Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C20orf203 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182584.2 | NP_872390.2 | 5 | 3192 | ||
GenBank mRNA | AK091025.1 | - | 5 | 3089 | ||
GU931819.1 | - | 6 | 3502 | |||
GU931820.1 | - | 6 | 3502 |