Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00319 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026960.1 | 3 | 2724 | |||
GenBank mRNA | AK095355.1 | - | 5 | 3164 | ||
AK128444.1 | - | 3 | 2724 | |||
BC128187.1 | - | 3 | 2662 | |||
BC128188.1 | - | 3 | 2349 |
Gene Symbol: CH507-42P11.6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006723919.2 | XP_006723982.1 | 2 | 3390 | ||
XM_011546086.1 | XP_011544388.1 | 2 | 3339 | |||
XM_017028528.1 | XP_016884017.1 | 2 | 2975 |