Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATXN8OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002717.2 | 4 | 851 | |||
GenBank mRNA | AF126749.1 | - | LIT3320 SCA8 |
851 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000424524.1 | - | 4 | 430 | Chr. 13 - 70107912 - 70131546 [+] on Build GRCh38 |