Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MROH7-TTC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037639.2 | 10, 11 | 2317 | |||
NR_037641.2 | 9, 10 | 2137 | ||||
GenBank mRNA | CR749830.1 | - | 10, 11 | 2269 |
Gene Symbol: MROH7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026782.2 | 9, 10 | 2131 | |||
GenBank mRNA | AK130047.1 | - | 8, 9 | 2626 | ||
BC095478.1 | - | 8, 9 | 1919 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AL045710.1 | - | 3, 4 | 221 | Chr. 1 - 54670909 - 54680010 [+] on Build GRCh38 |