Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEXA-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027262.1 | 1 | 1297 | |||
GenBank mRNA | AK023377.1 | - | 1 | 1045 | ||
BC068211.1 | - | 1 | 1297 |