Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPT-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024559.1 | 2 | 363 | |||
GenBank mRNA | AW770488.1 | - | 2 | 363 | ||
BM714794.1 | - | 1 | 269 |