Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM254-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027428.1 | 5 | 1033 | |||
NR_027429.1 | 4 | 946 | ||||
NR_027430.1 | 4 | 822 | ||||
NR_027431.2 | 4 | 796 | ||||
NR_027432.1 | 3 | 709 | ||||
GenBank mRNA | AK021673.1 | - | 4 | 665 | ||
AK096758.1 | - | 5 | 735 | |||
AK308811.1 | - | 3 | 262 | |||
AK309654.1 | - | 4 | 445 | |||
AK310182.1 | - | 4 | 498 | |||
AK310261.1 | - | 3 | 399 | |||
BC028046.1 | - | 3 | 370 |