Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA2P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024261.2 | 9 | 1839 | |||
GenBank mRNA | AL137680.1 | - | 9 | 1743 | ||
BC034587.1 | - | 1 | 530 |