Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028092.1 | 3 | 359 | |||
NR_028093.1 | 3 | 359 | ||||
GenBank mRNA | U19517.1 | - | 3 | 359 | ||
U19518.1 | - | 3 | 359 |