Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00052 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026869.1 | 3 | 524 | |||
GenBank mRNA | AK056023.1 | - | 3 | 523 | ||
BC066551.1 | - | 3 | 508 |