Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THUMPD3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027007.3 | 3 | 781 | |||
NR_132780.1 | 4 | 855 | ||||
GenBank mRNA | AK093640.1 | - | 2 | 1906 | ||
AL110136.1 | - | 3 | 777 | |||
BC044625.2 | - | 1 | 1892 | |||
BC052961.1 | - | 3 | 3153 | |||
DB567546.1 | - | 1 | 448 |