Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00680-GUSBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003660.2 | 5 | 537 | |||
NR_132997.1 | 7 | 796 | ||||
NR_132998.1 | 4 | 464 | ||||
GenBank mRNA | BC144584.1 | - | 5 | 473 | ||
BC144585.1 | - | 4 | 400 | |||
BM553931.1 | - | 7 | 796 | |||
DA269273.1 | - | 4 | 294 |
Gene Symbol: GUSBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132999.1 | 3 | 297 | |||
NR_133000.1 | 3 | 297 | ||||
GenBank mRNA | AW955360.1 | - | 2 | 125 |