Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024191.1 | 1, 2 | 148 | |||
XM_011533022.2 | XP_011531324.1 | 1, 2 | 143 | |||
GenBank mRNA | AK303985.1 | - | 1, 2 | 132 | ||
AK308894.1 | - | 1, 2 | 133 | |||
BC053508.1 | - | 1, 2 | 120 | 6338889 |