Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBPF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005271144.3 | XP_005271201.1 | 15 | 2815 | ||
XM_011542012.2 | XP_011540314.1 | 20 | 4144 | |||
XM_011542014.2 | XP_011540316.1 | 15 | 3453 | |||
XM_017002148.1 | XP_016857637.1 | 20 | 3279 |
Gene Symbol: NBPF22P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003719.2 | 8 | 1996 | |||
GenBank mRNA | BC050328.1 | - | 8 | 1952 |
Gene Symbol: NBPF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011540763.1 | XP_011539065.1 | 16 | 2764 |