Gene Symbol: HERC2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002824.3 | 16 | 2335 | |||
GenBank mRNA | AB002391.2 | - | 14 | 2080 | ||
AF041080.1 | - | LIT2190 HERC2P2 |
16 | 2339 | ||
AF041081.1 | - | LIT2191 HERC2P3 |
14 | 2015 | ||
CR749409.1 | - | 4 | 606 |
Gene Symbol: HERC2P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036432.1 | 15 | 2174 | |||
GenBank mRNA | AK294795.1 | - | 11 | 1720 | ||
BC000975.2 | - | 4 | 460 | |||
BC033888.1 | - | 15 | 2179 | |||
BC094757.1 | - | 15 | 2174 | |||
X69636.1 | - | 3 | 327 |
Gene Symbol: HERC2P9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BI007057.1 | - | 2 | 291 |