Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHACTR2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027113.2 | 3 | 645 | |||
NR_027114.2 | 3 | 1050 | ||||
NR_110148.1 | 5 | 845 | ||||
NR_110149.1 | 3 | 387 | ||||
GenBank mRNA | AK093151.1 | - | 3 | 1018 | ||
AK098520.1 | - | 3 | 645 | |||
BC033369.1 | - | 3 | 1050 | |||
HY000603.1 | - | 3 | 356 |