Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026873.1 | 5 | 4380 | |||
GenBank mRNA | AK091213.1 | - | 1 | 3038 | ||
AK092625.1 | - | 1 | 2157 | |||
AK124597.1 | - | 1 | 3501 | |||
BQ645787.1 | - | 1 | 393 |