Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPEAR-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026547.1 | 1 | 205 | |||
NR_026548.1 | 1 | 205 | ||||
GenBank mRNA | AF426269.1 | - | 1 | 205 | ||
AF426270.1 | - | 1 | 205 |