Gene Symbol: NIPSNAP3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018376.3 | NP_060846.2 | 6 | 5041 | ||
NR_130759.1 | 5 | 4830 | ||||
XM_011518839.1 | XP_011517141.1 | 5 | 4891 | |||
GenBank mRNA | AF370389.1 | - | 1 | 1329 | ||
AK090505.1 | - | 1 | 1900 | |||
AK091667.1 | - | 1 | 1334 |