Gene Symbol: NIPSNAP3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018376.3 | NP_060846.2 | 6 | 5324 | ||
NR_130759.1 | 5 | 5113 | ||||
NR_130760.1 | 6 | 1223 | ||||
XM_011518839.1 | XP_011517141.1 | 5 | 5174 | |||
XM_017014889.1 | XP_016870378.1 | 6 | 1145 | |||
GenBank mRNA | AF370389.1 | - | 1 | 1612 | ||
AK091667.1 | - | 1 | 1617 | |||
BX107655.1 | - | 2 | 122 |