Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC107133515 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015341.2 | 9 | 1389 | |||
GenBank mRNA | AK075282.1 | - | 5 | 753 | ||
BC118647.1 | - | 5 | 872 |
Gene Symbol: LRRC37BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124001.1 | - | 2 | 1127 | ||
AK124759.1 | - | 2 | 1534 | |||
AL713754.1 | - | 2 | 652 |