Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF438 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026560.1 | 5, 6 | 476 | |||
XM_017015875.1 | XP_016871364.1 | 6, 7 | 462 | |||
XM_017015876.1 | XP_016871365.1 | 8, 9 | 1211 | |||
XM_017015877.1 | XP_016871366.1 | 7, 8 | 641 | |||
XM_017015878.1 | XP_016871367.1 | 6, 7 | 568 | |||
XM_017015879.1 | XP_016871368.1 | 6, 7 | 648 | |||
XM_017015880.1 | XP_016871369.1 | 5, 6 | 451 | |||
XM_017015881.1 | XP_016871370.1 | 4, 5 | 389 | |||
XM_017015882.1 | XP_016871371.1 | 5, 6 | 496 | |||
GenBank mRNA | AK057323.1 | - | 5, 6 | 464 | ||
AK294253.1 | - | 6, 7 | 452 |