Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSUN5P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094312.1 | - | 1 | 1426 | ||
AK126486.1 | - | 3 | 3400 | |||
AK128374.1 | - | 1 | 1638 |