Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282275.1 | NP_001269204.1 | 1 | 134 | ||
GenBank mRNA | AF532777.1 | - | 1 | 122 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CB048981.1 | - | 1 | 98 | Chr. 17 - 48723209 - 48724729 [+] on Build GRCh38 |