Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTERF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028049.1 | 7 | 3827 | |||
NR_028050.1 | 5 | 2019 | ||||
NR_028051.1 | 4 | 1834 | ||||
XM_006712294.1 | XP_006712357.1 | 5 | 2288 | |||
XM_006712295.3 | XP_006712358.1 | 5 | 2444 | |||
XM_011510634.2 | XP_011508936.1 | 7 | 4130 | |||
XM_017003379.1 | XP_016858868.1 | 7 | 4758 | |||
XM_017003381.1 | XP_016858870.1 | 8 | 4594 | |||
XM_017003382.1 | XP_016858871.1 | 8 | 4107 | |||
XM_017003383.1 | XP_016858872.1 | 8 | 4880 | |||
XM_017003384.1 | XP_016858873.1 | 8 | 4855 | |||
XM_017003386.1 | XP_016858875.1 | 7 | 5229 | |||
XM_017003387.1 | XP_016858876.1 | 6 | 4781 | |||
XM_017003389.1 | XP_016858878.1 | 6 | 3541 | |||
XM_017003390.1 | XP_016858879.1 | 5 | 4014 | |||
XM_017003391.1 | XP_016858880.1 | 5 | 5034 | |||
GenBank mRNA | AK025374.1 | - | 1 | 1079 | ||
AK093695.1 | - | 5 | 1974 | |||
BC044641.1 | - | 1 | 587 | |||
BX648184.1 | - | 7 | 3792 |